1-Chloro-3,5-dimethoxybenzene ≥97.0%

Supplier: TCI America

Synonyms: 5-Chloro-1,3-dimethoxybenzene

C1258-25G C1258-10G
TCC1258-25GEA 477.16 CAD
TCC1258-25G TCC1258-010G
1-Chloro-3,5-dimethoxybenzene ≥97.0%
1-Chloro-3,5-dimethoxybenzene

CAS Number: 7051-16-3
MDL Number: MFCD00008382
Molecular Formula: C8H9ClO2
Molecular Weight: 172.61
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Boiling point (°C): 113
Melting point (°C): 35
Flash Point (°C): 112


Product Categories: Materials Chemistry, Dendrimer Building Blocks
Other Product Names / Synonyms: 5-Chloro-1,3-dimethoxybenzene

Formula: C₈H₉ClO₂
MW: 172.61 g/mol
Boiling Pt: 113 °C
Melting Pt: 35 °C
Flash Pt: 112 °C
MDL Number: MFCD00008382
CAS Number: 7051-16-3
EINECS: 230-330-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3e08c096574f38c56e70d5abdc5e5e8471826e60&vid=3e08c096574f38c56e70d5abdc5e5e8471826e60&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR