5-Chloro-2-hydroxypyridine ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 3-Chloro-6-pyridinol, 5-Chloro-2-pyridinol, 3-Chloro-6-hydroxypyridine

C1245-5G C1245-25G
TCC1245-005GEA 37.92 CAD
TCC1245-005G TCC1245-025G
5-Chloro-2-hydroxypyridine ≥98.0% (by GC, titration analysis)
5-Chloro-2-hydroxypyridine

CAS Number: 4214-79-3
MDL Number: MFCD00006275
Molecular Formula: C5H4ClNO
Molecular Weight: 129.54
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 164


Other Product Names / Synonyms: 5-Chloro-2-pyridinol, 5-Chloro-2(1H)-pyridinone, 5-Chloro-2-pyridone

Formula: C₅H₄ClNO
MW: 129.55 g/mol
Melting Pt: 163
MDL Number: MFCD00006275
CAS Number: 4214-79-3
EINECS: 224-146-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f91c49a2d7639b07a01806ea68b9c74040c6bba9&vid=12332f99788218add41b1b6ab6e9a9dcb208a5c0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR