4-Chlorophenyl methyl sulfone ≥98.0%

Supplier: TCI America

C1237-5G C1237-25G
TCC1237-5GEA 55.3 CAD
TCC1237-5G TCC1237-025G
4-Chlorophenyl methyl sulfone ≥98.0%
4-Chlorophenyl methyl sulfone

CAS Number: 98-57-7
MDL Number: MFCD00025067
Molecular Formula: C7H7ClO2S
Molecular Weight: 190.64
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 60
Melting point (°C): 98

Formula: C₇H₇ClO₂S
Boiling Pt: 60 °C
Melting Pt: 98 °C
MDL Number: MFCD00025067
CAS Number: 98-57-7
EINECS: 202-682-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d271e074133000f175af6a1ef203485519b1dcc5&vid=d271e074133000f175af6a1ef203485519b1dcc5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR