5-Chloro-2-thenoic acid ≥95.0% (by GC, titration analysis)

Supplier: TCI America

C1230-25G C1230-5G
TCC1230-25GEA 71.1 CAD
TCC1230-25G TCC1230-005G
5-Chloro-2-thenoic acid ≥95.0% (by GC, titration analysis)
5-Chloro-2-thenoic acid

CAS Number: 24065-33-6
MDL Number: MFCD00041426
Molecular Formula: C5H3ClO2S
Molecular Weight: 162.59
Purity/Analysis Method: >95.0% (GC,T)
Form: Crystal
Melting point (°C): 151

Formula: C₅H₃ClO₂S
Melting Pt: 151 °C
MDL Number: MFCD00041426
CAS Number: 24065-33-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=75a5bd9eb9d9d49b62e32c3f0fbe07b060cd6406&vid=75a5bd9eb9d9d49b62e32c3f0fbe07b060cd6406&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR