2-Chloro-5-nitrocinnamic acid ≥97.0%

Supplier: TCI America
Warning

Synonyms: 3-(2-Chloro-5-nitrophenyl)acrylic acid

C1206-5G C1206-25G
TCC1206-005GEA 137.46 CAD
TCC1206-005G TCC1206-025G
2-Chloro-5-nitrocinnamic acid ≥97.0%
2-Chloro-5-nitrocinnamic acid

CAS Number: 36015-19-7
MDL Number: MFCD00063312
Molecular Formula: C9H6ClNO4
Molecular Weight: 227.60
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 221

Formula: C₉H₆ClNO₄
MW: 227.6 g/mol
Melting Pt: 221 °C
MDL Number: MFCD00063312
CAS Number: 36015-19-7
UN: 3077
ADR: 9 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a648e50b9b42db8258adb527b0f6394655320cc3&vid=a648e50b9b42db8258adb527b0f6394655320cc3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR