4-Chloro-3-nitrocinnamic acid ≥97.0%

Supplier: TCI America

Synonyms: 3-(4-Chloro-3-nitrophenyl)prop-2-enoic acid, 3-(4-Chloro-3-nitrophenyl)acrylic acid

C1205-25G C1205-5G
TCC1205-25GEA 479.37 CAD
TCC1205-25G TCC1205-005G
4-Chloro-3-nitrocinnamic acid ≥97.0%
4-Chloro-3-nitrocinnamic acid

CAS Number: 20797-48-2
MDL Number: MFCD00063311
Molecular Formula: C9H6ClNO4
Molecular Weight: 227.60
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 187

Formula: C₉H₆ClNO₄
MW: 227.601 g/mol
Melting Pt: 187 °C
MDL Number: MFCD00063311
CAS Number: 20797-48-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=51792a89e8d2ad93f21cc0b5ed43d3c1f69707f8&vid=51792a89e8d2ad93f21cc0b5ed43d3c1f69707f8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR