2-Chloro-5-(trifluoromethyl)pyridine ≥97.0%
Supplier: TCI America
C1194-25G
C1194-100G
TCC1194-025GEA
64.78
CAD
TCC1194-025G
TCC1194-100G
2-Chloro-5-(trifluoromethyl)pyridine ≥97.0%
2-Chloro-5-(trifluoromethyl)pyridine
CAS Number: 52334-81-3
MDL Number: MFCD00042225
Molecular Formula: C6H3ClF3N
Molecular Weight: 181.54
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Boiling point (°C): 147
Flash Point (°C): 65
Freezing point (°C): 29
Formula:
C₆H₃ClF₃N Boiling Pt: 147 °C Melting Pt: 30…31 °C Density: 1.41 g/cm³ (20 °C) Flash Pt: 65 °C |
MDL Number:
MFCD00042225 CAS Number: 52334-81-3 EINECS: 257-856-5 UN: 2811 ADR: 6.1,III |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...