4-Chloro-2-fluoroaniline ≥98.0%

Supplier: TCI America

C1169-25G C1169-10G
TCC1169-025GEA 139.04 CAD
TCC1169-025G TCC1169-010G
4-Chloro-2-fluoroaniline ≥98.0%
4-Chloro-2-fluoroaniline

CAS Number: 57946-56-2
MDL Number: MFCD00010625
Molecular Formula: C6H5ClFN
Molecular Weight: 145.56
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 92
Flash Point (°C): 99
Specific Gravity (20/20): 1.33

Formula: C₆H₅ClFN
Boiling Pt: 92 °C
Density: 1.33 (20 °C)
Flash Pt: 99 °C
MDL Number: MFCD00010625
CAS Number: 57946-56-2
EINECS: 261-034-1
UN: 2810
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c038d250a556ea97075dc22a88052324485259dd&vid=5effc25faa3f84a5e82bdee76723e3e335a48b69&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR