4-Chloro-3-nitrobenzaldehyde ≥95.0% (by titrimetric analysis)

Supplier: TCI America

C1165-25G C1165-5G
TCC1165-025GEA 118.5 CAD
TCC1165-025G TCC1165-005G
4-Chloro-3-nitrobenzaldehyde ≥95.0% (by titrimetric analysis)
4-Chloro-3-nitrobenzaldehyde

CAS Number: 16588-34-4
MDL Number: MFCD00007078
Molecular Formula: C7H4ClNO3
Molecular Weight: 185.56
Purity/Analysis Method: >95.0% (T)
Form: Crystal
Melting point (°C): 62

Formula: C₇H₄ClNO₃
Melting Pt: 62 °C
Storage Temperature: Ambient
MDL Number: MFCD00007078
CAS Number: 16588-34-4
EINECS: 240-645-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8c25dce59cbdc3a623e35edf38598889f8de2fc0&vid=41ff19ffb008a73d80dac3e3c50cdec8b6820931&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR