2-Chloro-3-hydroxypyridine ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

C1152-10G C1152-25G
TCC1152-010GEA 63.2 CAD
TCC1152-010G TCC1152-25G
2-Chloro-3-hydroxypyridine ≥98.0% (by GC, titration analysis)
2-Chloro-3-hydroxypyridine

CAS Number: 6636-78-8
MDL Number: MFCD00006235
Molecular Formula: C5H4ClNO
Molecular Weight: 129.54
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 167


Other Product Names / Synonyms: 2-Chloro-3-pyridinol

Formula: C₅H₄ClNO
Boiling Pt: 212 °C (1013 hPa)
Melting Pt: 167 °C
MDL Number: MFCD00006235
CAS Number: 6636-78-8
EINECS: 229-635-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0ea5fc9371ae7ac50d80ddc5829cb8718e313c81&vid=0ea5fc9371ae7ac50d80ddc5829cb8718e313c81&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR