1,3-Cyclopentanedione ≥98.0%

Supplier: TCI America

C1146-1G C1146-5G
TCC1146-001GEA 58.46 CAD
TCC1146-001G TCC1146-005G
1,3-Cyclopentanedione ≥98.0%
1,3-Cyclopentanedione

CAS Number: 3859-41-4
MDL Number: MFCD00001405
Molecular Formula: C5H6O2
Molecular Weight: 98.10
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 150

Formula: C₅H₆O₂
Melting Pt: 150 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00001405
CAS Number: 3859-41-4
EINECS: 223-372-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=60ab7a21f43541481a3bdc040add09614e3aaf7e&vid=3342d82b77153721af907a726401f8d52ab774a9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR