2-Chlorobenzonitrile ≥98.0%

Supplier: TCI America
Warning

C1130-25G C1130-500G
TCC1130-025GEA 52.39 CAD
TCC1130-025G TCC1130-500G
2-Chlorobenzonitrile ≥98.0%
2-Chlorobenzonitrile

CAS Number: 873-32-5
MDL Number: MFCD00001779
Molecular Formula: C7H4ClN
Molecular Weight: 137.57
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 232
Melting point (°C): 45
Flash Point (°C): 108

Formula: C₇H₄ClN
Boiling Pt: 232 °C
Melting Pt: 45 °C
Density: 1.19 g/cm³ (25 °C)
Flash Pt: 108 °C
Storage Temperature: Ambient
MDL Number: MFCD00001779
CAS Number: 873-32-5
EINECS: 212-836-5
UN: 3439
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8a973a7836b267318a0d46da79369e1e31f679b7&vid=316d5143224004d4f17dc837955e9bf2aa0e958d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR