3-Chloro-4-hydroxybenzoic acid ≥98.0%

Supplier: TCI America
Warning

C1128-250G C1128-25G
TCC1128-250GEA 908.5 CAD
TCC1128-250G TCC1128-025G
3-Chloro-4-hydroxybenzoic acid ≥98.0%
3-Chloro-4-hydroxybenzoic acid

CAS Number: 3964-58-7
MDL Number: MFCD00002549
Molecular Formula: C7H5ClO3
Molecular Weight: 172.56
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 171

Formula: C₇H₅ClO₃
MW: 172.57 g/mol
Boiling Pt: 331
Melting Pt: 171 °C
MDL Number: MFCD00002549
CAS Number: 3964-58-7
EINECS: 223-574-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f4f199895b237da8744a62ad61ad4771b81be666&vid=f4f199895b237da8744a62ad61ad4771b81be666&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR