(2-Chloro-6-fluorophenyl)acetic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

C1126-25G C1126-5G
TCC1126-025GEA 168.59 CAD
TCC1126-025G TCC1126-5G
(2-Chloro-6-fluorophenyl)acetic acid ≥98.0% (by titrimetric analysis)
(2-Chloro-6-fluorophenyl)acetic acid

CAS Number: 37777-76-7
MDL Number: MFCD00004319
Molecular Formula: C8H6ClFO2
Molecular Weight: 188.58
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 123

Formula: (C₆H₃ClF)CH₂COOH
Boiling Pt: 289 °C (1013 hPa)
Melting Pt: 123 °C
Density: 1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004319
CAS Number: 37777-76-7
EINECS: 253-661-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a29803b9c0c360c731808081707f321dbd6bb2bd&vid=a29803b9c0c360c731808081707f321dbd6bb2bd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR