N-(Benzyloxycarbonyloxy)succinimide ≥98.0% (by total nitrogen basis)

Supplier: TCI America

Synonyms: CBZ-OSu (Z-OSu)

C1124-250G C1124-25G
TCC1124-250GEA 347.6 CAD
TCC1124-250G TCC1124-025G
N-(Benzyloxycarbonyloxy)succinimide ≥98.0% (by total nitrogen basis)
N-(Benzyloxycarbonyloxy)succinimide

CAS Number: 13139-17-8
MDL Number: MFCD00005513
Molecular Formula: C12H11NO5
Molecular Weight: 249.22
Purity/Analysis Method: >98.0% (N)
Form: Crystal
Melting point (°C): 81


Product Categories: Synthetic Organic Chemistry, Condensation & Active Esterification, Protection & Derivatization Reagents (for Synthesis), Peptide Synthesis, Bioscience, Polyamine Synthesis Reagents
Other Product Names / Synonyms: O-Cbz-N-hydroxysuccinimide, Benzyl N-Succinimidyl Carbonate, Cbz-OSu, Z-OSu

Formula: C₁₂H₁₁NO₅
MW: 249.22 g/mol
Melting Pt: 81 °C
MDL Number: MFCD00005513
CAS Number: 13139-17-8
EINECS: 236-075-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ea677ed6a44c876fd66e9c9ae32c0410972ff238&vid=ea677ed6a44c876fd66e9c9ae32c0410972ff238&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR