Methyl-4-cyanobenzoate ≥98.0%

Supplier: TCI America

C1119-250G C1119-25G
TCC1119-250GEA 690.46 CAD
TCC1119-250G TCC1119-025G
Methyl-4-cyanobenzoate ≥98.0%
Methyl 4-cyanobenzoate

CAS Number: 1129-35-7
MDL Number: MFCD00001823
Molecular Formula: C9H7NO2
Molecular Weight: 161.16
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 142
Melting point (°C): 68


Other Product Names / Synonyms: 4-Cyanobenzoic Acid Methyl Ester

Formula: C₉H₇NO₂
Boiling Pt: 142 °C
Melting Pt: 68 °C
MDL Number: MFCD00001823
CAS Number: 1129-35-7
EINECS: 214-443-4
UN: 3439
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7c7c08b9b9cff48fc9bdac80a3e41a76c75b012a&vid=7c7c08b9b9cff48fc9bdac80a3e41a76c75b012a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR