2-Chloronicotinic acid ≥98.0%

Supplier: TCI America
Warning

Synonyms: 2-Chloro-3-pyridinecarboxylic acid, 2-Chloropyridine-3-carboxylic acid

C1108-250G C1108-25G
TCC1108-250GEA 158 CAD
TCC1108-250G TCC1108-025G
2-Chloronicotinic acid ≥98.0%
2-Chloronicotinic acid

CAS Number: 2942-59-8
MDL Number: MFCD00006236
Molecular Formula: C6H4ClNO2
Molecular Weight: 157.55
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal


Other Product Names / Synonyms: 2-Chloro-3-pyridinecarboxylic Acid

Formula: C₆H₄ClNO₂
MW: 157.56 g/mol
Boiling Pt: 284
Melting Pt: 181
Storage Temperature: Ambient
MDL Number: MFCD00006236
CAS Number: 2942-59-8
EINECS: 220-937-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7b2fbe6cbdb0d2456e9798fe0a8b60d7196aa1a4&vid=7b2fbe6cbdb0d2456e9798fe0a8b60d7196aa1a4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR