2(5H)-Furanone ≥93.0%

Supplier: TCI America

C1078-5ML C1078-25ML
TCC1078-005MLEA 87.3 CAD
TCC1078-005ML TCC1078-025ML
2(5H)-Furanone ≥93.0%
2(5H)-Furanone

CAS Number: 497-23-4
MDL Number: MFCD00005376
Molecular Formula: C4H4O2
Molecular Weight: 84.07
Purity/Analysis Method: >93.0% (GC)
Form: Clear Liquid
Boiling point (°C): 214
Melting point (°C): 5
Flash Point (°C): 101
Specific Gravity (20/20): 1.20
Storage Temperature: 0-10°C


Product Categories: Bioscience, Lipids
Other Product Names / Synonyms: 2(5H)-Furanone, 2-Oxo-2,5-dihydrofuran

Formula: C₄H₄O₂
Boiling Pt: 214 °C
Melting Pt: 5 °C
Density: 1.20 (20 °C)
Flash Pt: 101 °C
MDL Number: MFCD00005376
CAS Number: 497-23-4
EINECS: 207-839-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eb0f46e14ced0838bc8d96990107f3ba86e31171&vid=eb0f46e14ced0838bc8d96990107f3ba86e31171&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 93.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR