2-(4-Chlorobenzoyl)benzoic acid ≥98.0%

Supplier: TCI America

C1074-25G C1074-500G
TCC1074-025GEA 28.44 CAD
TCC1074-025G TCC1074-500G
2-(4-Chlorobenzoyl)benzoic acid ≥98.0%
2-(4-Chlorobenzoyl)benzoic acid

CAS Number: 85-56-3
MDL Number: MFCD00002474
Molecular Formula: C14H9ClO3
Molecular Weight: 260.67
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 151


Other Product Names / Synonyms: 2-(4-Chlorobenzoyl)benzoic Acid

Formula: C₁₄H₉ClO₃
MW: 260.68 g/mol
Melting Pt: 151 °C
MDL Number: MFCD00002474
CAS Number: 85-56-3
EINECS: 201-615-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=552d729da4e4bc1b1385be09bac4a9701e1158ce&vid=552d729da4e4bc1b1385be09bac4a9701e1158ce&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR