(4-Carboxybutyl)triphenylphosphonium bromide ≥98.0%

Supplier: TCI America

Synonyms: (4-Carboxybut-1-yl)(triphenyl)phosphonium bromide

C1061-250G C1061-25G
TCC1061-250GEA 968.54 CAD
TCC1061-250G TCC1061-025G
(4-Carboxybutyl)triphenylphosphonium bromide ≥98.0%
(4-Carboxybutyl)triphenylphosphonium bromide

CAS Number: 17814-85-6
MDL Number: MFCD00011906
Molecular Formula: C23H24BrO2P
Molecular Weight: 443.32
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 207

Formula: C₂₃H₂₄BrO₂P
MW: 443.32 g/mol
Melting Pt: 207 °C
MDL Number: MFCD00011906
CAS Number: 17814-85-6
EINECS: 241-782-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d629b6738d690d0cc076820775025e0ffba531c5&vid=d629b6738d690d0cc076820775025e0ffba531c5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR