(3-Chlorophenyl)acetic acid 98,0

Supplier: TCI America
Warning

Synonyms: 3-Chlorophenylacetic acid

C1055-5G C1055-25G
TCC1055-005GEA 36.34 CAD
TCC1055-005G TCC1055-025G
(3-Chlorophenyl)acetic acid 98,0
(3-Chlorophenyl)acetic acid

CAS Number: 1878-65-5
MDL Number: MFCD00004332
Molecular Formula: C8H7ClO2
Molecular Weight: 170.59
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 81

Formula: (C₆H₄Cl)CH₂COOH
MW: 170.6 g/mol
Boiling Pt: 292
Melting Pt: 81 °C
Storage Temperature: Ambient
MDL Number: MFCD00004332
CAS Number: 1878-65-5
EINECS: 217-520-0
UN: 2811
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a627108335dde93e747b4a87477cfaab674ed9d6&vid=a627108335dde93e747b4a87477cfaab674ed9d6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR