3-Chloro-1-iodopropane ≥98.0% stabilized
Supplier: TCI America
|
Warning
|
Stabilization:
Stabilised with Stabilised
C1037-25G
C1037-5G
TCC1037-025GEA
222.78
CAD
TCC1037-025G
76219-660
3-Chloro-1-iodopropane ≥98.0% stabilized
3-Chloro-1-iodopropane
(stabilized with Copper chip)
CAS Number: 6940-76-7
MDL Number: MFCD00001096
Molecular Formula: C3H6ClI
Molecular Weight: 204.44
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 171
Flash Point (°C): 110
Specific Gravity (20/20): 1.95
Other Product Names / Synonyms: 3-Chloropropyl Iodide
Formula:
C₃H₆ClI Boiling Pt: 171 °C Density: 1.95 (20 °C) Flash Pt: 110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001096 CAS Number: 6940-76-7 EINECS: 230-088-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...