(1S)-(-)-Camphanic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

C1021-1G C1021-5G
TCC1021-001GEA 41.08 CAD
TCC1021-001G TCC1021-005G
(1S)-(-)-Camphanic acid ≥98.0% (by titrimetric analysis)
(1S)-(-)-Camphanic acid

CAS Number: 13429-83-9
MDL Number: MFCD00044948
Molecular Formula: C10H14O4
Molecular Weight: 198.22
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 201
Specific rotation [a]20/D: -18 deg (C=1, Dioxane)

Formula: C₁₀H₁₄O₄
Melting Pt: 201 °C
MDL Number: MFCD00044948
CAS Number: 13429-83-9
EINECS: 236-552-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e58513947cf970426eb939c9069e3b4399a8ea9b&vid=e58513947cf970426eb939c9069e3b4399a8ea9b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR