2-(Chloromethyl)quinoline hydrochloride ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
C1020-25G
C1020-250G
TCC1020-025GEA
173.8
CAD
TCC1020-025G
TCC1020-250G
2-(Chloromethyl)quinoline hydrochloride ≥98.0% (by HPLC, titration analysis)
2-(Chloromethyl)quinoline hydrochloride
CAS Number: 3747-74-8
MDL Number: MFCD00012734
Molecular Formula: C10H8ClN
Molecular Weight: 214.09
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 193
Formula:
C₁₀H₉Cl₂N Melting Pt: 193 °C |
MDL Number:
MFCD00012734 CAS Number: 3747-74-8 EINECS: 223-145-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...