2-(4-Chloro-3-nitrobenzoyl)benzoic acid ≥97.0%

Supplier: TCI America

C1013-25G C1013-250G
TCC1013-025GEA 75.84 CAD
TCC1013-025G TCC1013-250G
2-(4-Chloro-3-nitrobenzoyl)benzoic acid ≥97.0%
2-(4-Chloro-3-nitrobenzoyl)benzoic acid

CAS Number: 85-54-1
MDL Number: MFCD00007082
Molecular Formula: C14H8ClNO5
Molecular Weight: 305.67
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 195


Other Product Names / Synonyms: 4-Chloro-3-nitrobenzophenone-2'-carboxylic Acid

Formula: C₁₄H₈ClNO₅
MW: 305.67 g/mol
Melting Pt: 195 °C
MDL Number: MFCD00007082
CAS Number: 85-54-1
EINECS: 201-613-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=331ae1cab3c447c99d40f8eb5766eb2607a318f0&vid=331ae1cab3c447c99d40f8eb5766eb2607a318f0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR