Methyl carbamate ≥98.0%

Supplier: TCI America

Synonyms: Carbamic acid methyl ester

C1012-500G C1012-25G
TCC1012-500GEA 78.78 CAD
TCC1012-500G TCC1012-025G
Methyl carbamate ≥98.0%
Methyl carbamate

CAS Number: 598-55-0
MDL Number: MFCD00007964
Molecular Formula: C2H5NO2
Molecular Weight: 75.07
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 177
Melting point (°C): 54
Flash Point (°C): 100


Other Product Names / Synonyms: Carbamic Acid Methyl Ester

Formula: C₂H₅NO₂
MW: 75.07 g/mol
Boiling Pt: 177 °C
Melting Pt: 54 °C
Density: 1.1361 g/cm³ (56 °C)
Flash Pt: 100 °C
MDL Number: MFCD00007964
CAS Number: 598-55-0
EINECS: 209-939-2
Merck Index: 12,06115
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=32553508f1625a79a4ea3769d066076e064a4cf4&vid=32553508f1625a79a4ea3769d066076e064a4cf4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR