2-Chloro-3-nitropyridine ≥99.0%
Supplier: TCI America
|
Warning
|
C1008-5G
C1008-25G
TCC1008-005GEA
31.6
CAD
TCC1008-005G
TCC1008-025G
2-Chloro-3-nitropyridine ≥99.0%
2-Chloro-3-nitropyridine
CAS Number: 5470-18-8
MDL Number: MFCD00006232
Molecular Formula: C5H3ClN2O2
Molecular Weight: 158.54
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Melting point (°C): 103
Flash Point (°C): 185
Formula:
C₅H₃ClN₂O₂ MW: 158.54 g/mol Boiling Pt: 260 °C (1013 hPa) Melting Pt: 103 °C Flash Pt: 185 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006232 CAS Number: 5470-18-8 EINECS: 226-799-8 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...