Isopropyl cyanoacetate ≥97.0%

Supplier: TCI America

Synonyms: Isopropyl 2-cyanoacetate

C1001-25ML C1001-500ML
TCC1001-025MLEA 58.46 CAD
TCC1001-025ML TCC1001-500ML
Isopropyl cyanoacetate ≥97.0%
Isopropyl cyanoacetate

CAS Number: 13361-30-3
MDL Number: MFCD00019842
Molecular Formula: C6H9NO2
Molecular Weight: 127.14
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 206
Flash Point (°C): 99
Specific Gravity (20/20): 1.01


Other Product Names / Synonyms: Cyanoacetic Acid Isopropyl Ester

Formula: C₆H₉NO₂
MW: 127.14 g/mol
Boiling Pt: 206 °C
Density: 1.01 (20 °C)
Flash Pt: 99 °C
MDL Number: MFCD00019842
CAS Number: 13361-30-3
EINECS: 236-421-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6f119e069ce51c02523dc5d2fdfc2ef18ad4b552&vid=6f119e069ce51c02523dc5d2fdfc2ef18ad4b552&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR