Sodium chlorodifluoroacetate ≥99.0% (by titrimetric analysis)

Supplier: TCI America

C0991-500G C0991-25G C0991-100G
TCC0991-500GEA 410.03 CAD
TCC0991-500G TCC0991-025G TCC0991-100G
Sodium chlorodifluoroacetate ≥99.0% (by titrimetric analysis)
Sodium chlorodifluoroacetate

CAS Number: 1895-39-2
MDL Number: MFCD00064771
Molecular Formula: C2HClF2O2
Molecular Weight: 152.46
Purity/Analysis Method: >99.0% (T)
Form: Crystal


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks, Difluoromethylating Agents
Other Product Names / Synonyms: Chlorodifluoroacetic Acid Sodium Salt

Formula: C₂ClF₂NaO₂
Melting Pt: 193…195 °C
MDL Number: MFCD00064771
CAS Number: 1895-39-2
EINECS: 217-586-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1e78d7cba15ce3150ee64c57ed606f44072fdcdc&vid=1e78d7cba15ce3150ee64c57ed606f44072fdcdc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR