4-Chlorobenzhydrol ≥98.0%

Supplier: TCI America

C0989-25G C0989-500G
TCC0989-025GEA 52.14 CAD
TCC0989-025G TCC0989-500G
4-Chlorobenzhydrol ≥98.0%
4-Chlorobenzhydrol

CAS Number: 119-56-2
MDL Number: MFCD00004491
Molecular Formula: C13H11ClO
Molecular Weight: 218.68
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 151
Melting point (°C): 61


Other Product Names / Synonyms: 4-Chlorodiphenylcarbinol, 4-Chlorodiphenylmethanol

Formula: C₁₃H₁₁ClO
Boiling Pt: 151 °C
Melting Pt: 61 °C
MDL Number: MFCD00004491
CAS Number: 119-56-2
EINECS: 204-333-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=72d0891eb91f54b6c1d29dcbe88c9c73a10e0ff6&vid=72d0891eb91f54b6c1d29dcbe88c9c73a10e0ff6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR