Trimellitic anhydride ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 1,2,4-Benzenetricarboxylic anhydride, 1,2,4-Benzenetricarboxylic 1,2-anhydride

C0959-25G C0959-500G
TCC0959-025GEA 37.15 CAD
TCC0959-025G TCC0959-500G
Trimellitic anhydride ≥98.0% (by titrimetric analysis)
Trimellitic anhydride

CAS Number: 552-30-7
MDL Number: MFCD00005925
Molecular Formula: C9H4O5
Molecular Weight: 192.13
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Boiling point (°C): 245
Melting point (°C): 168
Flash Point (°C): 227


Other Product Names / Synonyms: 4-Carboxyphthalic Anhydride

Formula: C₉H₄O₅
MW: 192.13 g/mol
Boiling Pt: 245 °C
Melting Pt: 168 °C
Flash Pt: 227 °C
MDL Number: MFCD00005925
CAS Number: 552-30-7
EINECS: 209-008-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=be1d11f6081035741ce236788637fea4fd103824&vid=be1d11f6081035741ce236788637fea4fd103824&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR