1-(4-Morpholino)cyclohexene ≥97.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
N-(1-Cyclohexen-1-yl)morpholine, 1-Morpholinocyclohexene, 4-(1-Cyclohexen-1-yl)morpholine
C0956-25ML
C0956-500ML
TCC0956-025MLEA
102.7
CAD
TCC0956-025ML
TCC0956-500ML
1-(4-Morpholino)cyclohexene ≥97.0% (by GC, titration analysis)
1-(4-Morpholino)cyclohexene
CAS Number: 670-80-4
MDL Number: MFCD00006163
Molecular Formula: C10H17NO
Molecular Weight: 167.25
Purity/Analysis Method: >97.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 140
Flash Point (°C): 68
Specific Gravity (20/20): 1.02
Storage Temperature: 0-10°C
Other Product Names / Synonyms: N-(1-Cyclohexen-1-yl)morpholine
Formula:
C₁₀H₁₇NO MW: 167.25 g/mol Boiling Pt: 140 °C Density: 1.02 (20 °C) Flash Pt: 68 °C |
MDL Number:
MFCD00006163 CAS Number: 670-80-4 EINECS: 211-579-6 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...