CAPS (3-(cyclohexylamino)propanesulfonic acid) ≥98.0% (by titrimetric analysis)

Supplier: TCI America

C0921-250G C0921-25G
TCC0921-250GEA 307.37 CAD
TCC0921-250G TCC0921-025G
CAPS (3-(cyclohexylamino)propanesulfonic acid) ≥98.0% (by titrimetric analysis)
CAPS (3-(Cyclohexylamino)propanesulphonic acid)

[Good's buffer component for biological research]
CAS Number: 1135-40-6
MDL Number: MFCD00003837
Molecular Formula: C9H19NO3S
Molecular Weight: 221.32
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Flash Point (°C): 110


Product Categories: Bioscience, Good's Buffering Agents
Other Product Names / Synonyms: CAPS

Formula: C₆H₁₁NH(CH₂)₃SO₃H
Melting Pt: 324 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00003837
CAS Number: 1135-40-6
EINECS: 214-492-1
Merck Index: 13,01773
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3eb9ef3b36b6bc62c5c38161b01dbb90f35b8951&vid=891052e7cb36ced2d1ff863656d368cd923e6646&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR