(2-Chlorophenyl)acetic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

Synonyms: 2-Chlorophenylacetic acid

C0916-500G C0916-25G
TCC0916-500GEA 342.86 CAD
TCC0916-500G TCC0916-025G
(2-Chlorophenyl)acetic acid ≥98.0% (by titrimetric analysis)
(2-Chlorophenyl)acetic acid

CAS Number: 2444-36-2
MDL Number: MFCD00004317
Molecular Formula: C8H7ClO2
Molecular Weight: 170.59
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 95

Formula: (C₆H₄Cl)CH₂COOH
MW: 170.6 g/mol
Melting Pt: 95 °C
Storage Temperature: Ambient
MDL Number: MFCD00004317
CAS Number: 2444-36-2
EINECS: 219-482-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3b2201effeb7bf5f14ae7ea46b8ed2e64684ed57&vid=3b2201effeb7bf5f14ae7ea46b8ed2e64684ed57&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR