(4-Chlorophenyl)acetic acid 97,0

Supplier: TCI America
Warning

Synonyms: 2-(4-Chlorophenyl)acetic acid, p-Chlorophenylacetic Acid

C0914-25G C0914-500G
TCC0914-025GEA 72.68 CAD
TCC0914-025G TCC0914-500G
(4-Chlorophenyl)acetic acid 97,0
(4-Chlorophenyl)acetic acid

CAS Number: 1878-66-6
MDL Number: MFCD00004344
Molecular Formula: C8H7ClO2
Molecular Weight: 170.59
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 105

Formula: (C₆H₄Cl)CH₂COOH
MW: 170.6 g/mol
Boiling Pt: 292
Melting Pt: 105 °C
Density: 1.43 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004344
CAS Number: 1878-66-6
EINECS: 217-521-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bdcaff787dc25bbcaab598094c674e55778b4e9f&vid=bdcaff787dc25bbcaab598094c674e55778b4e9f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR