Cyclopentylacetic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI America
C0894-5G
C0894-25G
TCC0894-005GEA
39.5
CAD
TCC0894-005G
TCC0894-025G
Cyclopentylacetic acid ≥98.0% (by GC, titration analysis)
Cyclopentylacetic acid
CAS Number: 1123-00-8
MDL Number: MFCD00001387
Molecular Formula: C7H12O2
Molecular Weight: 128.17
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 231
Flash Point (°C): 109
Freezing point (°C): 14
Specific Gravity (20/20): 1.02
Other Product Names / Synonyms: Cyclopentaneacetic Acid
Formula:
C₇H₁₂O₂ Boiling Pt: 231 °C Density: 1.02 (20 °C) Flash Pt: 109 °C |
MDL Number:
MFCD00001387 CAS Number: 1123-00-8 EINECS: 214-368-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...