4-Chloro-2-nitrobenzoic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

C0879-25G C0879-500G
TCC0879-025GEA 105.86 CAD
TCC0879-025G TCC0879-500G
4-Chloro-2-nitrobenzoic acid ≥98.0% (by titrimetric analysis)
4-Chloro-2-nitrobenzoic acid

CAS Number: 6280-88-2
MDL Number: MFCD00007214
Molecular Formula: C7H4ClNO4
Molecular Weight: 201.56
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 145
Flash Point (°C): 100

Formula: C₇H₄ClNO₄
MW: 201.57 g/mol
Melting Pt: 145 °C
Flash Pt: 100 °C
Storage Temperature: Ambient
MDL Number: MFCD00007214
CAS Number: 6280-88-2
EINECS: 228-483-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=07619136c833ebcceb00479e9992cded7463dfcb&vid=6bdd9294de8610c0c97dce494373dc2c4fd3a835&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR