2-Chloro-5-nitrobenzoic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

C0877-500G C0877-25G
TCC0877-500GEA 270.18 CAD
TCC0877-500G TCC0877-025G
2-Chloro-5-nitrobenzoic acid ≥98.0% (by GC, titration analysis)
2-Chloro-5-nitrobenzoic acid

CAS Number: 2516-96-3
MDL Number: MFCD00007294
Molecular Formula: C7H4ClNO4
Molecular Weight: 201.56
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 165
Flash Point (°C): 100

Formula: C₇H₄ClNO₄
Melting Pt: 165 °C
Density: 0.4 g/cm³ (20 °C)
Flash Pt: 100 °C
Storage Temperature: Ambient
MDL Number: MFCD00007294
CAS Number: 2516-96-3
EINECS: 219-739-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b993c4b13f6b68a61133eb6fe6f150c8dd6375a8&vid=554e37283eafe338b4228b0fdd504f443b83a4d8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR