15-Crown-5 (1,4,7,10,13-pentaoxacyclopentadecane, lead ionophore V) ≥97.0%

Supplier: TCI America
Warning

C0859-25ML C0859-5ML
TCC0859-025MLEA 447.93 CAD
TCC0859-025ML TCC0859-005ML
15-Crown-5 (1,4,7,10,13-pentaoxacyclopentadecane, lead ionophore V) ≥97.0%
15-Crown-5 (1,4,7,10,13-Pentaoxacyclopentadecane, Lead ionophore V)

CAS Number: 33100-27-5
MDL Number: MFCD00005110
Molecular Formula: C10H20O5
Molecular Weight: 220.27
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 116
Melting point (°C): -20
Flash Point (°C): 113
Specific Gravity (20/20): 1.11


Product Categories: Materials Chemistry, Macrocycles for Host-Guest Chemistry
Other Product Names / Synonyms: 1,4,7,10,13-Pentaoxacyclopentadecane

Formula: C₁₀H₂₀O₅
Boiling Pt: 116 °C
Melting Pt: –20 °C
Density: 1.11 (20 °C)
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00005110
CAS Number: 33100-27-5
EINECS: 251-379-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3d46a493e3eb674a72986b6edb831d85d10973cc&vid=3d46a493e3eb674a72986b6edb831d85d10973cc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR