2-Chloroethylammonium chloride ≥98.0% (by total nitrogen and titration analysis)
Supplier: TCI America
C0851-500G
C0851-25G
C0851-100G
TCC0851-500GEA
270.18
CAD
TCC0851-500G
TCC0851-025G
TCC0851-100G
2-Chloroethylammonium chloride ≥98.0% (by total nitrogen and titration analysis)
2-Chloroethylammonium chloride
CAS Number: 870-24-6
MDL Number: MFCD00012887
Molecular Formula: C2H6ClN
Molecular Weight: 115.99
Purity/Analysis Method: >98.0% (N,T)
Form: Crystal
Melting point (°C): 147
Other Product Names / Synonyms: 2-Aminoethyl Chloride Hydrochloride
Formula:
C₂H₇Cl₂N Melting Pt: 147 °C |
MDL Number:
MFCD00012887 CAS Number: 870-24-6 EINECS: 212-793-2 UN: 3261 ADR: 8,III |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...