4-Hydroxybenzonitrile ≥98.0%

Supplier: TCI America
Warning

Synonyms: p-Cyanophenol, 4-Cyanophenol, p-Hydroxybenzonitrile

C0842-500G C0842-25G
TCC0842-500GEA 331.8 CAD
TCC0842-500G TCC0842-025G
4-Hydroxybenzonitrile ≥98.0%
4-Hydroxybenzonitrile

CAS Number: 767-00-0
MDL Number: MFCD00002312
Molecular Formula: C7H5NO
Molecular Weight: 119.12
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 146
Melting point (°C): 112


Product Categories: Materials Chemistry, Building Blocks for Liquid Crystals
Other Product Names / Synonyms: 4-Cyanophenol

Formula: C₇H₅NO
Boiling Pt: 146 °C
Melting Pt: 112 °C
Density: ∼1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002312
CAS Number: 767-00-0
EINECS: 212-175-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0106bd7722ffc7ce4219d4b8706abce6f6d83fda&vid=005b786f07e4b6e7b1b34cf05cea5e82083c3c08&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR