(3-Chloropropyl)trimethoxysilane ≥97.0%

Supplier: TCI America

Synonyms: 3-Chloropropyltrimethoxysilane, 3-(Trimethoxysilyl)propylchloride, Trimethoxy(3-chloropropyl)silane

C0840-25ML C0840-500ML C0840-100ML
TCC0840-025MLEA 34.76 CAD
TCC0840-025ML TCC0840-500ML TCC0840-100ML
(3-Chloropropyl)trimethoxysilane ≥97.0%
(3-Chloropropyl)trimethoxysilane

CAS Number: 2530-87-2
MDL Number: MFCD00000997
Molecular Formula: C6H15ClO3Si
Molecular Weight: 198.72
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 195
Melting point (°C): -50
Flash Point (°C): 78
Specific Gravity (20/20): 1.08


Product Categories: Materials Chemistry, Silane Coupling Agents
Other Product Names / Synonyms: 3-Chloropropyltrimethoxysilane

Formula: ClH₂CCH₂CH₂Si(OCH₃)₃
MW: 198.72 g/mol
Boiling Pt: 195
Melting Pt: -50
Density: 1.08 (20 °C)
Flash Pt: 78 °C
Storage Temperature: Ambient
MDL Number: MFCD00000997
CAS Number: 2530-87-2
EINECS: 219-787-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=df1c17342d13ea55ec4b3aefb4d92b628575cd8b&vid=df1c17342d13ea55ec4b3aefb4d92b628575cd8b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR