Cyclopentane-1,1-diacetic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

C0838-5G
TCC0838-005GEA 79 CAD
TCC0838-005G
Cyclopentane-1,1-diacetic acid ≥98.0% (by GC, titration analysis)
Cyclopentane-1,1-diacetic acid

CAS Number: 16713-66-9
MDL Number: MFCD00001388
Molecular Formula: C9H14O4
Molecular Weight: 186.21
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 177


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 3,3-Tetramethyleneglutaric Acid

Formula: C₉H₁₄O₄
Melting Pt: 177 °C
MDL Number: MFCD00001388
CAS Number: 16713-66-9
EINECS: 240-761-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0ed03ed1644ee31e73aa3f9220018a32cfc5477b&vid=0ed03ed1644ee31e73aa3f9220018a32cfc5477b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR