Cyclohexane ≥99.5%

Supplier: TCI America

Synonyms: HexahydrobenzeneHexamethylene

C0818-500ML
TCC0818-500MLEA 135.88 CAD
TCC0818-500ML
Cyclohexane ≥99.5%
Cyclohexane

[for HPLC Solvent]
CAS Number: 110-82-7
MDL Number: MFCD00003814
Molecular Formula: C6H12
Molecular Weight: 84.16
Purity/Analysis Method: >99.5% (GC)
Form: Clear Liquid
Boiling point (°C): 81
Flash Point (°C): -20
Freezing point (°C): 6
Specific Gravity (20/20): 0.78

Formula: C₆H₁₂
MW: 84.16 g/mol
Boiling Pt: 81 °C
Melting Pt: 7 °C
Density: 0.78 (20 °C)
Flash Pt: –20 °C
MDL Number: MFCD00003814
CAS Number: 110-82-7
UN: 1145
ADR: 3,II
Merck Index: 13,02752
https://ca.vwr.com/stibo/web/std.lang.all/78/57/28937857.jpg

Order Now

Specification Test Results

Purity/Analysis Method 99.5% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR