(±)-trans-1,2-Cyclohexanediol ≥99.0%

Supplier: TCI America

Synonyms: (±)-trans-Cyclohexane-1,2-diol, trans-Cyclohexane-1,2-diol

C0809-5G C0809-25G
TCC0809-005GEA 34.46 CAD
TCC0809-005G TCC0809-025G
(±)-trans-1,2-Cyclohexanediol ≥99.0%
(±)-trans-1,2-Cyclohexanediol

CAS Number: 1460-57-7
MDL Number: MFCD00063611
Molecular Formula: C6H12O2
Molecular Weight: 116.16
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 120
Melting point (°C): 102
Flash Point (°C): 134


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: trans-1,2-Dihydroxycyclohexane

Formula: C₆H₁₂O₂
Boiling Pt: 120 °C
Melting Pt: 102 °C
Density: 1.147 g/cm³ (24 °C)
Flash Pt: 134 °C
MDL Number: MFCD00063611
CAS Number: 1460-57-7
EINECS: 215-956-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a4661408dc78725004534d3c5d7de3cbb93780a7&vid=a4661408dc78725004534d3c5d7de3cbb93780a7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR