1,4-Cyclohexanedicarboxylic acid (mixture of cis and trans isomers) ≥98.0%

Supplier: TCI America

Synonyms: 1,4-Cyclohexanedicarboxylic acid

C0788-500G C0788-25G
TCC0788-500GEA 120.57 CAD
TCC0788-500G TCC0788-025G
1,4-Cyclohexanedicarboxylic acid (mixture of cis and trans isomers) ≥98.0%
1,4-Cyclohexanedicarboxylic acid (mixture of cis and trans isomers)

CAS Number: 1076-97-7
MDL Number: MFCD00001465
Molecular Formula: C8H12O4
Molecular Weight: 172.18
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Flash Point (°C): 235

Formula: C₈H₁₂O₄
MW: 172.18 g/mol
Flash Pt: 235 °C
MDL Number: MFCD00001465
CAS Number: 1076-97-7
EINECS: 214-068-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e7890872c6c6ad11d8830189a4083482c5c6c91d&vid=39c06fdc903583351db17bb4a69df1ac8cf328bb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR