2-Chloro-4-nitrobenzoic acid ≥98.0%

Supplier: TCI America
Warning

C0764-500G C0764-25G
TCC0764-500GEA 189.6 CAD
TCC0764-500G TCC0764-025G
2-Chloro-4-nitrobenzoic acid ≥98.0%
2-Chloro-4-nitrobenzoic acid

CAS Number: 99-60-5
MDL Number: MFCD00007209
Molecular Formula: C7H4ClNO4
Molecular Weight: 201.56
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 138

Formula: C₇H₄ClNO₄
MW: 201.57 g/mol
Melting Pt: 138 °C
Storage Temperature: Ambient
MDL Number: MFCD00007209
CAS Number: 99-60-5
EINECS: 202-771-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bc366b59bd73d3de5e622f66bfc8f2a198ff6fc5&vid=07cd6ceb939f7df10f21a25aa4dd81bcb3d01b54&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR