1-Chloroadamantane ≥98.0%

Supplier: TCI America

C0747-25G C0747-5G
TCC0747-025GEA 163.99 CAD
TCC0747-025G TCC0747-005G
1-Chloroadamantane ≥98.0%
1-Chloroadamantane

CAS Number: 935-56-8
MDL Number: MFCD00075627
Molecular Formula: C10H15Cl
Molecular Weight: 170.68
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 166


Other Product Names / Synonyms: 1-Adamantyl Chloride

Formula: C₁₀H₁₅Cl
Melting Pt: 166 °C
Density: 1.26 g/cm³ (20 °C)
MDL Number: MFCD00075627
CAS Number: 935-56-8
EINECS: 213-305-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cfdd45bc5ee2c4a8ef13a040ef22b8119637d851&vid=cfdd45bc5ee2c4a8ef13a040ef22b8119637d851&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR