5-Chlorosalicylaldehyde ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 5-Chloro-2-hydroxybenzaldehyde

C0738-25G C0738-5G
TCC0738-025GEA 83.74 CAD
TCC0738-025G TCC0738-5G
5-Chlorosalicylaldehyde ≥98.0% (by GC, titration analysis)
5-Chlorosalicylaldehyde

CAS Number: 635-93-8
MDL Number: MFCD00003331
Molecular Formula: C7H5ClO2
Molecular Weight: 156.57
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 101


Other Product Names / Synonyms: 5-Chloro-2-hydroxybenzaldehyde

Formula: C₇H₅ClO₂
MW: 156.57 g/mol
Boiling Pt: 105 °C (12 mmHg)
Melting Pt: 101 °C
MDL Number: MFCD00003331
CAS Number: 635-93-8
EINECS: 211-244-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0ea2244af79557388c0177d6aa81cd65d3080a82&vid=0ea2244af79557388c0177d6aa81cd65d3080a82&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR