1,4-Cyclohexanedione ≥98.0%

Supplier: TCI America

C0730-25G C0730-500G
TCC0730-025GEA 91.97 CAD
TCC0730-025G TCC0730-500G
1,4-Cyclohexanedione ≥98.0%
1,4-Cyclohexanedione

CAS Number: 637-88-7
MDL Number: MFCD00001606
Molecular Formula: C6H8O2
Molecular Weight: 112.13
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 132
Melting point (°C): 78
Flash Point (°C): 132

Formula: C₆H₈O₂
Boiling Pt: 132 °C
Melting Pt: 78 °C
Density: 1.28 g/cm³ (20 °C)
Flash Pt: 132 °C
Storage Temperature: Ambient
MDL Number: MFCD00001606
CAS Number: 637-88-7
EINECS: 211-306-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ab502d319dac7c003e436e735dc6e63be0fbb898&vid=77fbb6329712b4c5c73af6d8a73ae888889dbf05&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR